2-Chlor-5-(trifluormethyl)benzoesäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chlor-α,α,α-trifluor-m-toluylsäure

PC5584-25G PC5584-100G PC5584-5G
APOSPC5584-25GEA 29 EUR
APOSPC5584-25G APOSPC5584-100G APOSPC5584-5G
2-Chlor-5-(trifluormethyl)benzoesäure 96%
2-Chlor-5-(trifluormethyl)benzoesäure
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224.57 g/mol
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061094
CAS-Nummer: 657-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR