N-Methoxy-N-methyltrifluoroacetamide 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,2,2-Trifluoro-N-methoxy-N-methylacetamide
PC5591-25G
PC5591-100G
APOSPC5591-25GEA
118
EUR
APOSPC5591-25G
APOSPC5591-100G
N-Methoxy-N-methyltrifluoroacetamide 98%
N-Methoxy-N-methyltrifluoroacetamide
Formel:
C₄H₆F₃NO₂ Molecular Weight: 157,09 g/mol Siedepunkt: 52 °C/30mmHg Dichte: 1,3 g/cm³ (25 °C) Flammpunkt: 47,8 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077565 CAS-Nummer: 104863-67-4 UN: 1992 ADR: 3+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...