Methyl-2-bromo-3,3,3-trifluoropropanoate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-bromo-3,3,3-trifluoropropanoate

PC5595-5G PC5595-1G PC5595-250MG
APOSPC5595-5GEA 618 EUR
APOSPC5595-5G APOSPC5595-1G APOSPC5595-250MG
Methyl-2-bromo-3,3,3-trifluoropropanoate 97%
Methyl-2-bromo-3,3,3-trifluoropropanoate
Formel: C₄H₄BrF₃O₂
Molecular Weight: 220,9726 g/mol
Siedepunkt: 126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038319
CAS-Nummer: 113816-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR