Pentafluoriodethan 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Perfluorethyliodid, Iodpentafluorethan

PC5600-100G PC5600-25G
APOSPC5600-100GEA 374 EUR
APOSPC5600-100G APOSPC5600-25G
Pentafluoriodethan 98%
Pentafluoriodethan

Reagent for introduction of pentafluoroethyl groups into organic molecules: J.O.C., 1987,52, 2481.

Formel: C₂F₅I
Molecular Weight: 245.92 g/mol
Siedepunkt: 41609 °C
Schmelzpunkt: -92
Dichte: 2.07 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039400
CAS-Nummer: 354-64-3
EINECS: 206-566-7
UN: 3163
ADR: 2.2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR