Pentafluoroethyl phenyl ketone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3,3-Pentafluoropropiophenone, Pentafluoroethyl phenyl ketone

PC5605-25G
APOSPC5605-25GEA 495 EUR
APOSPC5605-25G
Pentafluoroethyl phenyl ketone 97%
Pentafluoroethyl phenyl ketone
Formel: C₉H₅F₅O
Molecular Weight: 224,13 g/mol
Siedepunkt: 159…161 °C
Dichte: 1,372 g/cm³ (25 °C)
Flammpunkt: 63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00018062
CAS-Nummer: 394-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR