4'-Brom-2',5'-difluoracetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2,5-difluorophenyl)ethanone

PC56074-1G PC56074-250MG PC56074-5G
APOSPC56074-1GEA 24.8 EUR
APOSPC56074-1G APOSPC56074-250MG APOSPC56074-5G
4'-Brom-2',5'-difluoracetophenon
4'-Brom-2',5'-difluoracetophenon
Formel: C₈H₅BrF₂O
Molecular Weight: 235,0255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526896
CAS-Nummer: 123942-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR