1-Nitro-2,4-bis-(trifluoromethyl)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Bis(trifluoromethyl)nitrobenzene

PC56080-1G PC56080-5G PC56080-250MG
APOSPC56080-1GEA 343 EUR
APOSPC56080-1G APOSPC56080-5G APOSPC56080-250MG
1-Nitro-2,4-bis-(trifluoromethyl)benzene
1-Nitro-2,4-bis-(trifluoromethyl)benzene
Formel: C₈H₃F₆NO₂
Molecular Weight: 259.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185347
CAS-Nummer: 224044-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR