5-Fluor-2-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific
Gefahr

PC5609-1G PC5609-500MG PC5609-5G PC5609-25G
APOSPC5609-1GEA 21.9 EUR
APOSPC5609-1G APOSPC5609-500MG APOSPC5609-5G APOSPC5609-25G
5-Fluor-2-(trifluormethyl)benzonitril
5-Fluor-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Molecular Weight: 189,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236281
CAS-Nummer: 240800-45-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR