Fluoroiodomethane 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Fluoromethyl iodide

PC56130-5G PC56130-1G
APOSPC56130-5GEA 475 EUR
APOSPC56130-5G APOSPC56130-1G
Fluoroiodomethane 95%
Fluoroiodomethane
Formel: CH₂FI
Molecular Weight: 159.93 g/mol
Siedepunkt: 54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12404952
CAS-Nummer: 373-53-5
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR