5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid
PC5614-1G
PC5614-250MG
APOSPC5614-1GEA
19
EUR
APOSPC5614-1G
APOSPC5614-250MG
5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid 97%
5'-Trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-carboxylic acid
Formel:
C₁₂H₁₃F₃N₂O₂ Schmelzpunkt: 162…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209248 CAS-Nummer: 406476-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...