2-Amino-4-(4-fluorphenyl)thiazol

Lieferant: Apollo Scientific
Achtung

PC5617-5G PC5617-25G
APOSPC5617-5GEA 63.8 EUR
APOSPC5617-5G APOSPC5617-25G
2-Amino-4-(4-fluorphenyl)thiazol
2-Amino-4-(4-fluorophenyl)thiazole
Formel: C₉H₇FN₂S
Molecular Weight: 194.2287 g/mol
Schmelzpunkt: 102…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01086598
CAS-Nummer: 77815-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR