1-Fluor-2-(trifluormethylsulfonyl)benzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Fluoro-2-((trifluoromethyl)sulphonyl)benzene
PC56254-1G
PC56254-5G
PC56254-10G
PC56254-25G
APOSPC56254-1GEA
21.9
EUR
APOSPC56254-1G
APOSPC56254-5G
APOSPC56254-10G
APOSPC56254-25G
1-Fluor-2-(trifluormethylsulfonyl)benzol
1-Fluor-2-(trifluormethylsulfonyl)benzol
Formel:
C₇H₄F₄O₂S Molecular Weight: 228.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09971596 CAS-Nummer: 2358-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...