2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC5628-1G
PC5628-25G
PC5628-5G
APOSPC5628-1GEA
36.4
EUR
APOSPC5628-1G
APOSPC5628-25G
APOSPC5628-5G
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate 95%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate
Formel:
C₁₄H₁₄F₈O₄ Molecular Weight: 398,25 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00078359 CAS-Nummer: 66818-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...