2,3,6-Trifluorisonicotinsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2,3,6-Trifluoropyridine-4-carboxylic acid

PC5635-10G PC5635-1G PC5635-5G
APOSPC5635-10GEA 841 EUR
APOSPC5635-10G APOSPC5635-1G APOSPC5635-5G
2,3,6-Trifluorisonicotinsäure 97%
2,3,6-Trifluorisonicotinsäure
Formel: C₆H₂F₃NO₂
Molecular Weight: 177.08 g/mol
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840204
CAS-Nummer: 675602-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR