2,3,5-Trifluorbenzoylchlorid 98%

Lieferant: Apollo Scientific
Gefahr

PC5643-10G PC5643-5G PC5643-1G
APOSPC5643-10GEA 237 EUR
APOSPC5643-10G APOSPC5643-5G APOSPC5643-1G
2,3,5-Trifluorbenzoylchlorid 98%
2,3,5-Trifluorbenzoylchlorid
Formel: C₇H₂ClF₃O
Molecular Weight: 194,54 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 240800-48-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR