2,3,4-Trifluorbenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,4-Trifluorbenzolsulfonsäureamid

PC5647-10G PC5647-1G PC5647-5G
APOSPC5647-10GEA 763 EUR
APOSPC5647-10G APOSPC5647-1G APOSPC5647-5G
2,3,4-Trifluorbenzolsulfonamid
2,3,4-Trifluorbenzolsulfonamid
Formel: C₆H₄F₃NO₂S
Molecular Weight: 211,16 g/mol
Schmelzpunkt: 110…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094300
CAS-Nummer: 518070-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR