4-Fluorphenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

PC5659-100G
APOSPC5659-100GEA 88.6 EUR
APOSPC5659-100G
4-Fluorphenylboronsäurepinakolester 97%
4-Fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₆BFO₂
Molecular Weight: 222,0637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03453665
CAS-Nummer: 214360-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR