Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid) 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (Chloro)(perfluorophenyl)dimethylsilane

PC5664-25G PC5664-5G
APOSPC5664-25GEA 106 EUR
APOSPC5664-25G APOSPC5664-5G
Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid) 98%
Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid)
Formel: C₈H₆ClF₅Si
Molecular Weight: 260,67 g/mol
Siedepunkt: 88…90 °C/10mmHg
Dichte: 1,381 g/cm³ (25 °C)
Flammpunkt: 95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000500
CAS-Nummer: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR