3,5-Difluorthiophenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluorbenzolthiol, 3,5-Difluorphenylmercaptan, 1,3-Difluor-5-mercaptobenzol, 3,5-Difluorbenzol-1-thiol

PC5677-1G PC5677-25G PC5677-5G
APOSPC5677-1GEA 23.2 EUR
APOSPC5677-1G APOSPC5677-25G APOSPC5677-5G
3,5-Difluorthiophenol
3,5-Difluorthiophenol
Formel: C₆H₄F₂S
Molecular Weight: 146,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06201795
CAS-Nummer: 99389-26-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR