2,3-Difluor-5-methylbenzonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC56801-1G
PC56801-500MG
APOSPC56801-1GEA
551
EUR
APOSPC56801-1G
APOSPC56801-500MG
2,3-Difluor-5-methylbenzonitril 95%
2,3-Difluor-5-methylbenzonitril
Formel:
C₈H₅F₂N Lagertemperatur: Kühlschrank |
MDL:
MFCD09996953 CAS-Nummer: 1003712-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...