3-Chloro-6-(trifluoromethyl)pyridazine

Lieferant: Apollo Scientific
Achtung

PC5684-1G PC5684-250MG
APOSPC5684-1GEA 58 EUR
APOSPC5684-1G APOSPC5684-250MG
3-Chloro-6-(trifluoromethyl)pyridazine
3-Chloro-6-(trifluoromethyl)pyridazine
Formel: C₅H₂ClF₃N₂
Molecular Weight: 182.53 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275188
CAS-Nummer: 258506-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR