3-Amino-6-fluor-1-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

PC5691-1G PC5691-250MG
APOSPC5691-1GEA 335 EUR
APOSPC5691-1G APOSPC5691-250MG
3-Amino-6-fluor-1-methyl-1H-indazol
3-Amino-6-fluoro-1-methyl-1H-indazole
Formel: C₈H₈FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11109398
CAS-Nummer: 171809-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR