3-(Pentafluorophenyl)propionic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Perfluorophenyl)propanoic acid
PC5700-1G
PC5700-5G
APOSPC5700-1GEA
183
EUR
APOSPC5700-1G
APOSPC5700-5G
3-(Pentafluorophenyl)propionic acid
3-(Pentafluorophenyl)propionic acid
Formel:
C₉H₅F₅O₂ Molecular Weight: 240,13 g/mol Schmelzpunkt: 91…96 °C Flammpunkt: 107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020516 CAS-Nummer: 2002-92-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...