Methyl-6-bromo-5-fluoronicotinate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-bromo-5-fluoronicotinate

PC57014-1G PC57014-5G PC57014-10G
APOSPC57014-1GEA 52.2 EUR
APOSPC57014-1G APOSPC57014-5G APOSPC57014-10G
Methyl-6-bromo-5-fluoronicotinate 95%
Methyl-6-bromo-5-fluoronicotinate
Formel: C₇H₅BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14698110
CAS-Nummer: 1214336-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR