2-Fluor-5-nitrobenzylalkohol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Fluoro-5-nitrophenyl)methanol
PC5704-10G
PC5704-5G
APOSPC5704-10GEA
152
EUR
APOSPC5704-10G
APOSPC5704-5G
2-Fluor-5-nitrobenzylalkohol 95+%
2-Fluor-5-nitrobenzylalkohol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,1300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683502 CAS-Nummer: 63878-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...