2-Bromo-4-chlorobenzotrifluoride
Lieferant: Apollo Scientific
|
Achtung
|
PC57042-25G
PC57042-5G
PC57042-100G
APOSPC57042-25GEA
92.8
EUR
APOSPC57042-25G
APOSPC57042-5G
APOSPC57042-100G
2-Bromo-4-chlorobenzotrifluoride
2-Bromo-4-chlorobenzotrifluoride
Formel:
C₇H₃BrClF₃ Molecular Weight: 259.46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839109 CAS-Nummer: 1099597-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...