5-Fluor-1H-indazol

Lieferant: Apollo Scientific
Achtung

PC5711-1G PC5711-250MG
APOSPC5711-1GEA 29 EUR
APOSPC5711-1G APOSPC5711-250MG
5-Fluor-1H-indazol
5-Fluoro-1H-indazole
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
Schmelzpunkt: 119…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972877
CAS-Nummer: 348-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR