5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC5716-100MG
PC5716-1G
PC5716-250MG
APOSPC5716-100MGEA
219
EUR
APOSPC5716-100MG
APOSPC5716-1G
APOSPC5716-250MG
5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester 95%
5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester
Formel:
C₁₄H₁₇BFNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026087 CAS-Nummer: 1150561-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...