2-(Trifluormethyl)-4-pyridinboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Trifluoromethyl)pyridin-4-yl)boronic acid

PC5751-1G PC5751-250MG PC5751-5G
APOSPC5751-1GEA 169 EUR
APOSPC5751-1G APOSPC5751-250MG APOSPC5751-5G
2-(Trifluormethyl)-4-pyridinboronsäure 95%
2-(Trifluoromethyl)-4-pyridineboronic acid
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190.9156 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11616522
CAS-Nummer: 1093407-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR