2-(4-Fluorobenzoyl)-3-(dimethylamino)acrylonitrile 95%
Lieferant: Apollo Scientific
PC5774-1G
PC5774-250MG
APOSPC5774-1GEA
429
EUR
APOSPC5774-1G
APOSPC5774-250MG
2-(4-Fluorobenzoyl)-3-(dimethylamino)acrylonitrile 95%
2-(4-Fluorobenzoyl)-3-(dimethylamino)acrylonitrile
Formel:
C₁₂H₁₁FN₂O Molecular Weight: 218,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03453098 CAS-Nummer: 52200-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...