Methyl-2,2,3,3,4,4-hexafluorobutanoate

Lieferant: Apollo Scientific

PC5781-10G PC5781-1G PC5781-5G
APOSPC5781-10GEA 864 EUR
APOSPC5781-10G APOSPC5781-1G APOSPC5781-5G
Methyl-2,2,3,3,4,4-hexafluorobutanoate
Methyl-2,2,3,3,4,4-hexafluorobutanoate
Formel: C₅H₄F₆O₂
Molecular Weight: 210,08 g/mol
Siedepunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00465645
CAS-Nummer: 356-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR