1,1,1,2,2-Pentafluor-3-iodpropan 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Iodo-1,1,1,2,2-pentafluoropropane

PC5787-1G PC5787-5G
APOSPC5787-1GEA 50.9 EUR
APOSPC5787-1G APOSPC5787-5G
1,1,1,2,2-Pentafluor-3-iodpropan 97%
1,1,1,2,2-Pentafluor-3-iodpropan
Formel: C₃H₂F₅I
Molecular Weight: 259,95 g/mol
Siedepunkt: 70…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00039405
CAS-Nummer: 354-69-8
EINECS: 206-568-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR