2-Brom-4-(trifluormethyl)phenylessigsäure

Lieferant: Apollo Scientific
Achtung

PC5830-1G PC5830-25G PC5830-5G
APOSPC5830-1GEA 60.9 EUR
APOSPC5830-1G APOSPC5830-25G APOSPC5830-5G
2-Brom-4-(trifluormethyl)phenylessigsäure
2-Brom-4-(trifluormethyl)phenylessigsäure
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 518070-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR