2-Fluoro-6-nitrobenzaldehyde 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC5841-1G
PC5841-5G
APOSPC5841-1GEA
33.5
EUR
APOSPC5841-1G
APOSPC5841-5G
2-Fluoro-6-nitrobenzaldehyde 99%
2-Fluoro-6-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ Schmelzpunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835320 CAS-Nummer: 1644-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...