Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride 90+%

Lieferant: Apollo Scientific
Gefahr

PC5843-5G PC5843-1G
APOSPC5843-5GEA 212 EUR
APOSPC5843-5G APOSPC5843-1G
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride 90+%
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoyl fluoride

Typically 92%, bal. other HFPO oligomer acid fluorides

Formel: C₁₈F₃₆O₆
Siedepunkt: 231…235 °C
Dichte: 1,796 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00054661
CAS-Nummer: 13252-15-8
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR