2-Brom-4,5-difluoranisol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4,5-difluoro-2-methoxybenzene
PC5863-25G
PC5863-5G
PC5863-10G
APOSPC5863-25GEA
58
EUR
APOSPC5863-25G
APOSPC5863-5G
APOSPC5863-10G
2-Brom-4,5-difluoranisol 97%
2-Brom-4,5-difluoranisol
Formel:
C₇H₅BrF₂O Schmelzpunkt: 23…25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070753 CAS-Nummer: 202865-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...