2,6-Difluor-4-iodanilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5886-25G
PC5886-5G
PC5886-100G
APOSPC5886-25GEA
60.9
EUR
APOSPC5886-25G
APOSPC5886-5G
APOSPC5886-100G
2,6-Difluor-4-iodanilin 98%
2,6-Difluor-4-iodanilin
Formel:
C₆H₄F₂IN Molecular Weight: 255,0039 g/mol Schmelzpunkt: 77…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774189 CAS-Nummer: 141743-49-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...