2-Fluoro-5-(trifluoromethoxy)benzamide 98%

Lieferant: Apollo Scientific

PC5898-5G PC5898-1G
APOSPC5898-5GEA 148 EUR
APOSPC5898-5G APOSPC5898-1G
2-Fluoro-5-(trifluoromethoxy)benzamide 98%
2-Fluoro-5-(trifluoromethoxy)benzamide
Formel: C₈H₅F₄NO₂
Molecular Weight: 223.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115896
CAS-Nummer: 886498-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR