2-Fluor-5-(trifluormethoxy)benzylamin

Lieferant: Apollo Scientific
Gefahr

PC5899-10G PC5899-1G PC5899-5G
APOSPC5899-10GEA 642 EUR
APOSPC5899-10G APOSPC5899-1G APOSPC5899-5G
2-Fluor-5-(trifluormethoxy)benzylamin
2-Fluor-5-(trifluormethoxy)benzylamin
Formel: C₈H₇F₄NO
Molecular Weight: 209,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115895
CAS-Nummer: 886498-13-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR