2-Fluor-p-toluonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5912-100G
PC5912-25G
PC5912-5G
APOSPC5912-100GEA
244
EUR
APOSPC5912-100G
APOSPC5912-25G
APOSPC5912-5G
2-Fluor-p-toluonitril 98%
2-Fluor-p-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol Schmelzpunkt: 52…53 °C Flammpunkt: 80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094311 CAS-Nummer: 85070-67-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...