5-Fluor-m-toluonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC5914-1G
PC5914-25G
PC5914-5G
PC5914-100G
APOSPC5914-1GEA
14.7
EUR
APOSPC5914-1G
APOSPC5914-25G
APOSPC5914-5G
APOSPC5914-100G
5-Fluor-m-toluonitril 95%
5-Fluor-m-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094312 CAS-Nummer: 216976-30-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...