3-Fluor-5-methylbenzaldehyd 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluor-m-tolualdehyd, 5-Fluor-m-tolylaldehyd, 3-Fluor-5-tolualdehyd

PC5917-1G PC5917-5G
APOSPC5917-1GEA 37.7 EUR
APOSPC5917-1G APOSPC5917-5G
3-Fluor-5-methylbenzaldehyd 99%
3-Fluoro-5-methylbenzaldehyde
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094315
CAS-Nummer: 189628-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR