3-Fluor-5-methylbenzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC5918-1G PC5918-25G PC5918-5G
APOSPC5918-1GEA 55.1 EUR
APOSPC5918-1G APOSPC5918-25G APOSPC5918-5G
3-Fluor-5-methylbenzylbromid 98%
3-Fluor-5-methylbenzylbromid
Formel: C₈H₈BrF
Lagertemperatur: Raumtemperatur
MDL: MFCD03094316
CAS-Nummer: 212268-39-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR