2-Fluor-4-methylbenzylbromid
Lieferant: Apollo Scientific
|
Gefahr
|
PC5936-10G
PC5936-1G
PC5936-5G
APOSPC5936-10GEA
364
EUR
APOSPC5936-10G
APOSPC5936-1G
APOSPC5936-5G
2-Fluor-4-methylbenzylbromid
2-Fluor-4-methylbenzylbromid
Formel:
C₈H₈BrF Molecular Weight: 203,05 g/mol Siedepunkt: 97…101 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094327 CAS-Nummer: 118745-63-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...