(2-Fluor-4-methylphenyl)essigsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-methylphenylacetic acid

PC5939-1G PC5939-5G PC5939-10G
APOSPC5939-1GEA 23.2 EUR
APOSPC5939-1G APOSPC5939-5G APOSPC5939-10G
(2-Fluor-4-methylphenyl)essigsäure 95%
(2-Fluor-4-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Molecular Weight: 168.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094329
CAS-Nummer: 518070-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR