2'-Fluor-4'-methylacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluoro-4-methylphenyl)ethanone

PC5942-1G PC5942-5G
APOSPC5942-1GEA 30.6 EUR
APOSPC5942-1G APOSPC5942-5G
2'-Fluor-4'-methylacetophenon
2'-Fluor-4'-methylacetophenon
Formel: C₉H₉FO
Molecular Weight: 152.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094331
CAS-Nummer: 29427-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR