1-Ethynyl-4-(trifluoromethoxy)benzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethoxy)phenylacetylene
PC5952-10G
PC5952-1G
PC5952-5G
APOSPC5952-10GEA
309
EUR
APOSPC5952-10G
APOSPC5952-1G
APOSPC5952-5G
1-Ethynyl-4-(trifluoromethoxy)benzene
1-Ethynyl-4-(trifluoromethoxy)benzene
Formel:
C₉H₅F₃O Molecular Weight: 186,13 g/mol Dichte: 1,215 g/cm³ (25 °C) Flammpunkt: 40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094333 CAS-Nummer: 160542-02-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...