(2,5-Bis(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

PC5973-1G PC5973-5G
APOSPC5973-1GEA 174 EUR
APOSPC5973-1G APOSPC5973-5G
(2,5-Bis(trifluormethyl)phenyl)essigsäure 98%
(2,5-Bis(trifluormethyl)phenyl)essigsäure
Formel: (C₆H₃(CF₃)₂)OCH₂COOH
Molecular Weight: 272,15 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 302912-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR