Bis(2-methoxyethyl)aminosulphur trifluoride 50% in THF
Lieferant: Apollo Scientific
Gefahr
PC6001-100G PC6001-10G PC6001-25G
APOSPC6001-100GEA 348 EUR
APOSPC6001-100G APOSPC6001-10G APOSPC6001-25G
Bis(2-methoxyethyl)aminosulphur trifluoride 50% in THF
Bis(2-methoxyethyl)aminosulphur trifluoride
Deoxo-FluorR (Registered trademark of Air Products), 50% solution in THF. Stable alternative to DAST.
Formel: C₆H₁₄F₃NO₂S
Molecular Weight: 221.24 g/mol
Dichte: 1.02 g/cm³ (25 °C)
Flammpunkt: <10 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01321415
CAS-Nummer: 202289-38-1
UN: 3286
ADR: 3+6.1+8 II