Perfluoro(2-ethoxyethane)sulphonic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Perfluoro(3-oxapentane-1-sulphonic acid)
PC6005-25G
PC6005-5G
APOSPC6005-25GEA
575
EUR
APOSPC6005-25G
APOSPC6005-5G
Perfluoro(2-ethoxyethane)sulphonic acid 97%
Perfluoro(2-ethoxyethane)sulphonic acid
Formel:
C₄F₉HO₄S Siedepunkt: 198 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00153237 CAS-Nummer: 113507-82-7 UN: 2586 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...